Learn More
Biolegend Alexa Fluor 700 anti-mouse CD4 Antibody, 100 ug
Supplier:  Biolegend 116022
Encompass_Preferred
CD4 is a 55 kD protein, also known as L3T4 or T4. It is a member of the Ig superfamily, primarily expressed on most thymocytes and a subset of T cells, and weakly on macrophages and dendritic cells. It acts as a coreceptor with the TCR during T cell activation and thymic differentiation by binding MHC class II and associating with the protein tyrosin kinase, lck.
Catalog No. NC1893173