Learn More
Biolegend Alexa Fluor 647 anti-mouse CD45
Supplier:  Biolegend 160303
Encompass_Preferred
Alexa Fluor 647 anti-mouse CD45 [S18009D]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC, IHC-F; Size: 25 ug
Catalog No. 50-207-2449