Learn More
Cell Signaling Technology Alexa Fluor™ 647 Phalloidin, Lyophilized, 300 Units
SDP
Catalog No. 502050592
Encompass
Alexa Fluor™ 647 Phalloidin allows researchers to fluorescently stain the cytoskeleton through the binding of phalloidin to F-actin. This product is designed for use on fixed and permeabilized samples. Reconstituted in anhydrous DMSO, the stock solution provides enough material to perform 200 assays at a 1:100 dilution and 100 μl assay volume.
  • Excitation and emission wavelengths of 650 nm and 668 nm, respectively.
  • Provided as a lyophilized solid stable for 1 year at -20°C when stored desiccated and protected from light.
  • Validated for immunofluorescence applications including frozen and immunocytochemistry samples.
  • High affinity binding to F-actin for cytoskeletal visualization.
  • Compatible with all species.
Catalog No. 50-205-0592 Supplier Cell Signaling Technology Supplier No. 8940S
Alexa Fluor™ 647 Phalloidin allows researchers to fluorescently stain the cytoskeleton through the binding of phalloidin to F-actin. This product is designed for use on fixed and permeabilized samples. Reconstituted in anhydrous DMSO, the stock solution provides enough material to perform 200 assays at a 1:100 dilution and 100 μl assay volume.
  • Excitation and emission wavelengths of 650 nm and 668 nm, respectively.
  • Provided as a lyophilized solid stable for 1 year at -20°C when stored desiccated and protected from light.
  • Validated for immunofluorescence applications including frozen and immunocytochemistry samples.
  • High affinity binding to F-actin for cytoskeletal visualization.
  • Compatible with all species.