Learn More
Abcam Ale x a Fluor 594 Conjugation Kit (Fast) - Lightning-Link, 1MG
Catalog No. NC2445719
Encompass_Preferred
Alexa Fluor 594 conjugation / labeling in < 20 mins with 30 secs hands-on time using Alexa Fluor 594 Conjugation Kit (Fast) - Lightning-Link ab269822. 100% antibody recovery.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC2445719 Supplier Abcam Supplier No. AB2698221MG
Alexa Fluor 594 conjugation / labeling in < 20 mins with 30 secs hands-on time using Alexa Fluor 594 Conjugation Kit (Fast) - Lightning-Link ab269822. 100% antibody recovery.
The product is subject to the following: Abcam Restricted Use Statement