Learn More
Biolegend Alexa Fluor 700 anti-human CD38
Supplier:  Biolegend 356623
Encompass_Preferred
Alexa Fluor 700 anti-human CD38 [HB-7]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 25 tests
Catalog No. NC1121543