Learn More
Biolegend Alexa Fluor 488 anti-α Smooth Muscle Actin [1A4]; Isotype: Mouse IgG2a, κ; Reactivity: Human, Mouse; Apps: IHC-P, ICC; Size: 100 μg
Catalog No. 502723718
Encompass_Preferred
Alexa Fluor 488 anti-α Smooth Muscle Actin [1A4]; Isotype: Mouse IgG2a, κ; Reactivity: Human, Mouse; Apps: IHC-P, ICC; Size: 100 μg
Catalog No. 50-272-3718 Supplier Biolegend Supplier No. 614854
Alexa Fluor 488 anti-α Smooth Muscle Actin [1A4]; Isotype: Mouse IgG2a, κ; Reactivity: Human, Mouse; Apps: IHC-P, ICC; Size: 100 μg