Learn More
Biolegend Alexa FluorR 488 anti-Cytochro
Catalog No. 502721820
Encompass_Preferred
Alexa Fluor 488 anti-Cytochrome c [6H2.B4]; Isotype: Mouse IgG1, κ; Reactivity: Mouse, Rat, Human; Apps: ICFC; Size: 100 μg
Catalog No. 50-272-1820 Supplier Biolegend Supplier No. 612308
Alexa Fluor 488 anti-Cytochrome c [6H2.B4]; Isotype: Mouse IgG1, κ; Reactivity: Mouse, Rat, Human; Apps: ICFC; Size: 100 μg