Learn More
Biolegend Alexa FluorR 488 anti-human CD
Supplier:  Biolegend 336232
Encompass_Preferred
Alexa Fluor 488 anti-human CD36 [5-271]; Isotype: Mouse IgG2a, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Catalog No. 50-272-2284