Learn More
Biolegend Alexa FluorR 488 anti-RBFOX3 N
Catalog No. 502721822
Encompass_Preferred
Alexa Fluor 488 anti-RBFOX3 (NeuN) [W18230A]; Isotype: Rat IgG2a, κ; Reactivity: Human, Mouse; Apps: IHC-F, IHC-P; Size: 100 μg
Catalog No. 50-272-1822 Supplier Biolegend Supplier No. 608456
Alexa Fluor 488 anti-RBFOX3 (NeuN) [W18230A]; Isotype: Rat IgG2a, κ; Reactivity: Human, Mouse; Apps: IHC-F, IHC-P; Size: 100 μg