Learn More
Biolegend Alexa FluorR 488 anti-TMEM119
Catalog No. 502740606
Encompass_Preferred
Alexa Fluor 488 anti-TMEM119 (Extracellular) [A16075D]; Isotype: Mouse IgG2b, κ; Reactivity: Human; Apps: ICFC, ICC; Size: 100 μg
Catalog No. 50-274-0606 Supplier Biolegend Supplier No. 853318
Alexa Fluor 488 anti-TMEM119 (Extracellular) [A16075D]; Isotype: Mouse IgG2b, κ; Reactivity: Human; Apps: ICFC, ICC; Size: 100 μg