Learn More
Biolegend Alexa FluorR 594 anti-Neurofil
Catalog No. 502721657
Encompass_Preferred
Alexa Fluor 594 anti-Neurofilament L (NF-L) [NFL3]; Isotype: Mouse IgG1, κ; Reactivity: Human, Mouse, Rat; Apps: IHC-P; Size: 25 μg
Catalog No. 50-272-1657 Supplier Biolegend Supplier No. 845905
Alexa Fluor 594 anti-Neurofilament L (NF-L) [NFL3]; Isotype: Mouse IgG1, κ; Reactivity: Human, Mouse, Rat; Apps: IHC-P; Size: 25 μg