Learn More
Biolegend Alexa FluorR 594 anti-Tyrosine
Catalog No. 502721665
Encompass_Preferred
Alexa Fluor 594 anti-Tyrosine Hydroxylase [2/40/15]; Isotype: Mouse IgG2a, κ; Reactivity: Human, Mouse, Rat; Apps: IHC-P; Size: 100 μg
Catalog No. 50-272-1665 Supplier Biolegend Supplier No. 818004
Alexa Fluor 594 anti-Tyrosine Hydroxylase [2/40/15]; Isotype: Mouse IgG2a, κ; Reactivity: Human, Mouse, Rat; Apps: IHC-P; Size: 100 μg