Learn More
Biolegend Alexa FluorR 647 anti-AMPKalph
Catalog No. 502740625
Encompass_Preferred
Alexa Fluor 647 anti-AMPKα Phospho (Thr172) [A20017A]; Isotype: Mouse IgG1, κ; Reactivity: Human, Mouse; Apps: IHC-P, ICFC; Size: 100 μg
Catalog No. 50-274-0625 Supplier Biolegend Supplier No. 600656
Alexa Fluor 647 anti-AMPKα Phospho (Thr172) [A20017A]; Isotype: Mouse IgG1, κ; Reactivity: Human, Mouse; Apps: IHC-P, ICFC; Size: 100 μg