Learn More
Biolegend Alexa Fluor 647 anti-CD340 (ErbB2/HER-2) [A20145B]; Isotype: Mouse IgG2b, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Catalog No. 502567073
Encompass_Preferred
Alexa Fluor 647 anti-CD340 (ErbB2/HER-2) [A20145B]; Isotype: Mouse IgG2b, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Catalog No. 50-256-7073 Supplier Biolegend Supplier No. 616154
Alexa Fluor 647 anti-CD340 (ErbB2/HER-2) [A20145B]; Isotype: Mouse IgG2b, κ; Reactivity: Human; Apps: FC; Size: 100 tests