Learn More
Biolegend Alexa FluorR 647 anti-mouse CD
Supplier:  Biolegend 114205
Encompass_Preferred
Alexa Fluor 647 anti-mouse CD326 (Ep-CAM) [W21115A]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: IHC-F, IHC-P; Size: 25 μg
Catalog No. 50-279-6454