Learn More
Biolegend Alexa FluorR 647 anti-Ubiquiti
Catalog No. 502721660
Encompass_Preferred
Alexa Fluor 647 anti-Ubiquitin [P4G7]; Isotype: Mouse IgG1, κ; Reactivity: Human, Mouse, Rat, Drosophila; Apps: IHC-P; Size: 100 μg
Catalog No. 50-272-1660 Supplier Biolegend Supplier No. 838710
Alexa Fluor 647 anti-Ubiquitin [P4G7]; Isotype: Mouse IgG1, κ; Reactivity: Human, Mouse, Rat, Drosophila; Apps: IHC-P; Size: 100 μg