Learn More
Biolegend Alexa Fluor 700 anti-human CD39 [A1]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cynomolgus, Rhesus; Apps: FC; Size: 25 tests
Supplier:  Biolegend 328255
Encompass_Preferred
Alexa Fluor 700 anti-human CD39 [A1]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cynomolgus, Rhesus; Apps: FC; Size: 25 tests
Catalog No. 50-272-3835