Learn More
Biolegend Alexa FluorR 700 anti-mouse CD
Catalog No. 502722642
Encompass_Preferred
Alexa Fluor 700 anti-mouse CD45 [I3/2.3]; Isotype: Rat IgG2b, λ; Reactivity: Mouse; Apps: FC; Size: 25 μg
Catalog No. 50-272-2642 Supplier Biolegend Supplier No. 147715
Alexa Fluor 700 anti-mouse CD45 [I3/2.3]; Isotype: Rat IgG2b, λ; Reactivity: Mouse; Apps: FC; Size: 25 μg