Learn More
Biolegend Alexa FluorR 700 anti-mouse IL
Catalog No. 502721889
Encompass_Preferred
Alexa Fluor 700 anti-mouse IL-12/IL-23 p40 [C15.6]; Isotype: Rat IgG1, κ; Reactivity: Mouse; Apps: ICFC; Size: 25 μg
Catalog No. 50-272-1889 Supplier Biolegend Supplier No. 505213
Alexa Fluor 700 anti-mouse IL-12/IL-23 p40 [C15.6]; Isotype: Rat IgG1, κ; Reactivity: Mouse; Apps: ICFC; Size: 25 μg