Learn More
BANGS LABORATORIES INC Quantum™ Alexa Fluor 647 MESF, Flow Cytometry Standardization, Accurate Calibration Curves, 1 mL
Catalog No. NC0334245
Encompass
The Quantum™ Alexa Fluor 647 MESF is a precise reference standard from Bangs Laboratories designed for fluorescence intensity calibration in flow cytometry. Its microbeads, mimicking human lymphocyte size, facilitate establishing calibration curves for accurate fluorescence measurements. The kit, including 5 distinct microsphere populations, enhances instrument linearity and detection thresholds, setting it apart for reliable quantitative analysis in cytometry.
Catalog No. NC0334245 Supplier BANGS LABORATORIES INC Supplier No. 647A1ML
The Quantum™ Alexa Fluor 647 MESF is a precise reference standard from Bangs Laboratories designed for fluorescence intensity calibration in flow cytometry. Its microbeads, mimicking human lymphocyte size, facilitate establishing calibration curves for accurate fluorescence measurements. The kit, including 5 distinct microsphere populations, enhances instrument linearity and detection thresholds, setting it apart for reliable quantitative analysis in cytometry.