Learn More
Jackson Immuno Research Labs Alexa Fluor 647 AffiniPure Donkey Anti-Goat IgG (H+L), .5mg
SDP
Catalog No. NC0299922
Encompass_Preferred
Alexa Fluor 647-AffiniPure Donkey Anti-Goat IgG (H+L) (min X Ck,GP,Sy Hms,Hrs,Hu,Ms,Rb,Rat Sr Prot)
Catalog No. NC0299922 Supplier Jackson Immuno Research Labs Supplier No. 705605147
Alexa Fluor 647-AffiniPure Donkey Anti-Goat IgG (H+L) (min X Ck,GP,Sy Hms,Hrs,Hu,Ms,Rb,Rat Sr Prot)