Learn More
Biolegend Alexa Fluor 647 anti-human CD68
Supplier:  Biolegend 333819
Encompass_Preferred
Alexa Fluor 647 anti-human CD68 [Y1/82A]; Isotype: Mouse IgG2b, κ; Reactivity: Human; Apps: ICFC; Size: 25 tests
Catalog No. NC1248367