Learn More
Apexbio Technology LLC Alexidine dihydrochloride 1715-30-6 10mM (in 1mL DMSO)
SDP
Catalog No. 5000061882
Encompass_Preferred
Alexidine dihydrochloride is a selective inhibitor targeting mitochondrial protein tyrosine phosphatase 1 (PTPMT1) exhibiting inhibitory activity with an IC50 value of approximately 1 08 M in vitro By inhibiting PTPMT1 Alexidine dihydrochloride modulates mitochondrial signaling pathways implicated in cellular metabolism Additionally this compound enhances insulin secretion in pancreatic -cells of rat islet models In cancer biology research Alexidine dihydrochloride demonstrates activity against FaDu cells providing a relevant tool for studying mitochondrial phosphatase-mediated mechanisms in oncology and diabetes-related research applications
Catalog No. 50-000-61882 Supplier Apexbio Technology LLC Supplier No. A45425.1
Alexidine dihydrochloride is a selective inhibitor targeting mitochondrial protein tyrosine phosphatase 1 (PTPMT1) exhibiting inhibitory activity with an IC50 value of approximately 1 08 M in vitro By inhibiting PTPMT1 Alexidine dihydrochloride modulates mitochondrial signaling pathways implicated in cellular metabolism Additionally this compound enhances insulin secretion in pancreatic -cells of rat islet models In cancer biology research Alexidine dihydrochloride demonstrates activity against FaDu cells providing a relevant tool for studying mitochondrial phosphatase-mediated mechanisms in oncology and diabetes-related research applications