Learn More
Medchemexpress LLC HY-108547 50mg Medchemexpress, Alexidine (dihydrochloride) CAS:1715-30-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY10854750MG
Encompass_Preferred
Medchemexpress, HY-108547 50mg Alexidine (dihydrochloride) CAS:1715-30-6 Alexidine dihydrochloride is an anticancer agent that targets a mitochondrial tyrosine phosphatase, PTPMT1, in mammalian cells and causes mitochondrial apoptosis. Alexidine dihydrochloride has antifungal and antibiofilm activity against a diverse range of fungal pathogens. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9636