Learn More
Medchemexpress LLC Alexidine dihydrochloride | 1715-30-6 | ≥98% | 581.71 | 1 ML
SDP
Catalog No. 5000346905
Encompass_Preferred
Alexidine dihydrochloride is an anticancer agent that targets a mitochondrial tyrosine phosphatase, PTPMT1, in mammalian cells, leading to mitochondrial apoptosis. It also exhibits broad-spectrum antifungal and antibiofilm activity against various fungal pathogens, including fluconazole-resistant strains and mature biofilms.
  • Targets mitochondrial tyrosine phosphatase (PTPMT1).
  • Induces mitochondrial apoptosis.
  • Shows broad-spectrum antifungal activity against diverse fungal pathogens.
  • Effective against fluconazole-resistant Candida isolates.
  • Inhibits filamentation and proliferation of fungi.
  • Decimates mature biofilms of Candida, Cryptococcus, and Aspergillus species at low concentrations.
  • Inhibits lateral yeast formation and biofilm dispersal in C. albicans.
  • Reduces fungal biofilm growth and viability by 67% in C. albicans murine models.
Catalog No. 50-003-46905 Supplier Medchemexpress LLC Supplier No. HY10854710MM/1ML
Alexidine dihydrochloride is an anticancer agent that targets a mitochondrial tyrosine phosphatase, PTPMT1, in mammalian cells, leading to mitochondrial apoptosis. It also exhibits broad-spectrum antifungal and antibiofilm activity against various fungal pathogens, including fluconazole-resistant strains and mature biofilms.
  • Targets mitochondrial tyrosine phosphatase (PTPMT1).
  • Induces mitochondrial apoptosis.
  • Shows broad-spectrum antifungal activity against diverse fungal pathogens.
  • Effective against fluconazole-resistant Candida isolates.
  • Inhibits filamentation and proliferation of fungi.
  • Decimates mature biofilms of Candida, Cryptococcus, and Aspergillus species at low concentrations.
  • Inhibits lateral yeast formation and biofilm dispersal in C. albicans.
  • Reduces fungal biofilm growth and viability by 67% in C. albicans murine models.