Learn More
Medchemexpress LLC Alexidine dihydrochloride | 1715-30-6 | 99.7% | 581.71 | 10 MG
SDP
Catalog No. 5000346904
Encompass_Preferred
Alexidine dihydrochloride is an anticancer agent that targets a mitochondrial tyrosine phosphatase, PTPMT1, in mammalian cells and causes mitochondrial apoptosis. It also has antifungal and antibiofilm activity against a diverse range of fungal pathogens.
  • Anticancer agent targeting mitochondrial tyrosine phosphatase, PTPMT1.
  • Causes mitochondrial apoptosis.
  • Antifungal and antibiofilm activity against a diverse range of fungal pathogens.
  • Displays activity against most Candida spp., including fluconazole-resistant isolates.
  • Complete inhibition of filamentation or proliferation of imaged fungi.
  • Able to decimate mature biofilms of Candida, Cryptococcus, and Aspergillus spp. at low concentrations.
  • Inhibits fungal biofilm growth and viability in a murine biofilm model.
Catalog No. 50-003-46904 Supplier Medchemexpress LLC Supplier No. HY10854710MG
Alexidine dihydrochloride is an anticancer agent that targets a mitochondrial tyrosine phosphatase, PTPMT1, in mammalian cells and causes mitochondrial apoptosis. It also has antifungal and antibiofilm activity against a diverse range of fungal pathogens.
  • Anticancer agent targeting mitochondrial tyrosine phosphatase, PTPMT1.
  • Causes mitochondrial apoptosis.
  • Antifungal and antibiofilm activity against a diverse range of fungal pathogens.
  • Displays activity against most Candida spp., including fluconazole-resistant isolates.
  • Complete inhibition of filamentation or proliferation of imaged fungi.
  • Able to decimate mature biofilms of Candida, Cryptococcus, and Aspergillus spp. at low concentrations.
  • Inhibits fungal biofilm growth and viability in a murine biofilm model.