Learn More
Medchemexpress LLC Alexidine (dihydrochloride) | 1715-30-6 | 99.7% | 581.71 | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHY108547
Encompass_Preferred
Alexidine dihydrochloride is an anticancer agent that targets a mitochondrial tyrosine phosphatase, PTPMT1, in mammalian cells and causes mitochondrial apoptosis. It also exhibits antifungal and antibiofilm activity against a diverse range of fungal pathogens.
  • Anticancer agent targeting PTPMT1.
  • Causes mitochondrial apoptosis.
  • Exhibits antifungal and antibiofilm activity against diverse fungal pathogens.
  • Effective against most Candida spp. with low MIC values.
  • Inhibits filamentation and proliferation of imaged fungi.
  • Decimates mature biofilms of Candida, Cryptococcus, and Aspergillus spp. at low concentrations.
  • Inhibits lateral yeast formation and biofilm dispersal.
  • Significantly reduces the density of C. albicans biofilms in jugular vein catheters of mice.
Catalog No. 50-003-47750