Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Alexidine dihydrochloride | 1715-30-6 | 99.7% | 581.71 g/mol | C26H58Cl2N10 | 5 MG
SDP

Catalog No. 5000263802
Encompass_Preferred

Alexidine dihydrochloride is a bis-biguanide dihydrochloride salt used in biochemical and microbiological research as a mitochondrial tyrosine phosphatase (PTPMT1) inhibitor and as an antifungal/antibiofilm agent. It is supplied as a solid with high purity and is suitable for in vitro studies and for preparation of in vivo dosing formulations.

  • Antifungal and antibiofilm activity demonstrated in diverse fungal pathogens.
  • Targets mitochondrial tyrosine phosphatase PTPMT1 and can induce mitochondrial apoptosis.
  • High purity (99.68%).
  • Soluble in DMSO (125 mg/mL) and compatible with several in vivo formulation protocols.
  • Stable when stored sealed at -20°C; solvent stability at -80°C for extended periods.
  • Molecular weight 581.71 g/mol; molecular formula C26H58Cl2N10.

Catalog No. 50-002-63802 Supplier Medchemexpress LLC Supplier No. HY1085475MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Alexidine dihydrochloride is a bis-biguanide dihydrochloride salt used in biochemical and microbiological research as a mitochondrial tyrosine phosphatase (PTPMT1) inhibitor and as an antifungal/antibiofilm agent. It is supplied as a solid with high purity and is suitable for in vitro studies and for preparation of in vivo dosing formulations.

  • Antifungal and antibiofilm activity demonstrated in diverse fungal pathogens.
  • Targets mitochondrial tyrosine phosphatase PTPMT1 and can induce mitochondrial apoptosis.
  • High purity (99.68%).
  • Soluble in DMSO (125 mg/mL) and compatible with several in vivo formulation protocols.
  • Stable when stored sealed at -20°C; solvent stability at -80°C for extended periods.
  • Molecular weight 581.71 g/mol; molecular formula C26H58Cl2N10.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.