Learn More
Selleck Chemical LLC Alexidine dihydrochloride-25MG
SDP
Catalog No. 503138982
Encompass_Preferred
Alexidine dihydrochloride a dibiguanide compound is an effective and selective inhibitor of protein tyrosine phosphatases localized to mitochondrion 1 (PTPMT1) with an IC50 of 1 08 M in vitro It has the potential to treat type II diabetes
Catalog No. 50-313-8982 Supplier Selleck Chemical LLC Supplier No. E178025MG
Alexidine dihydrochloride a dibiguanide compound is an effective and selective inhibitor of protein tyrosine phosphatases localized to mitochondrion 1 (PTPMT1) with an IC50 of 1 08 M in vitro It has the potential to treat type II diabetes