Learn More
TARGETMOL CHEMICALS INC Alexidine dihydrochloride
SDP
Catalog No. 502875871
Encompass_Preferred
Also available in 1 mL, 25 mg, 50 mg and bulk. Please contact Fisher for quotes. Alexidine dihydrochloride has antifungal and antibiofilm activity against a diverse range of fungal pathogens. Alexidine dihydrochloride is an anticancer agent that targets a mitochondrial tyrosine phosphatase, PTPMT1, in mammalian cells and causes mitochondrial apoptosis. Thus, Alexidine dihydrochloride has the potential to be developed as a pan-antifungal, antibiofilm drug. Purity 98.31%
Catalog No. 50-287-5871 Supplier TARGETMOL CHEMICALS INC Supplier No. T14177100MG
Also available in 1 mL, 25 mg, 50 mg and bulk. Please contact Fisher for quotes. Alexidine dihydrochloride has antifungal and antibiofilm activity against a diverse range of fungal pathogens. Alexidine dihydrochloride is an anticancer agent that targets a mitochondrial tyrosine phosphatase, PTPMT1, in mammalian cells and causes mitochondrial apoptosis. Thus, Alexidine dihydrochloride has the potential to be developed as a pan-antifungal, antibiofilm drug. Purity 98.31%