Learn More
Apexbio Technology LLC Alexidine dihydrochloride,50mg CAS# 1715-30-6
SDP
Supplier:  Apexbio Technology LLC A454250
Encompass_Preferred
Selective inhibitor of protein tyrosine phosphatases localized to mitochondrion 1 (PTPMT1) (IC50 = 1.08 μM in vitro). Stimulates increased insulin secretion by β-cells in rat pancreatic islets. Displays antitcancer properties in FaDu cells. Other sizes are also available. Please inqury us for quote.
Catalog No. 50-224-0163