Learn More
Cayman Chemical Alexidine (hydrochloride), Purity: ≥98%, CAS: 1715-30-6, 50 mg
SDP
Catalog No. 502954252
Encompass_Preferred
An alkyl bis(biguanide) antiseptic which kills plaque forming microorganisms; binds to LPS and LTA and inhibits fungal phospholipase B (IC50 ~ 250 nM); inhibits the mitochondrial phosphatase PTPMT1 (IC50 = 1.08 μM, in vitro) and induces apoptosis in cancer cell lines (ED50 = 1.8-2.6 μM)
Catalog No. 50-295-4252 Supplier Cayman Chemical Supplier No. 1387650 MG
An alkyl bis(biguanide) antiseptic which kills plaque forming microorganisms; binds to LPS and LTA and inhibits fungal phospholipase B (IC50 ~ 250 nM); inhibits the mitochondrial phosphatase PTPMT1 (IC50 = 1.08 μM, in vitro) and induces apoptosis in cancer cell lines (ED50 = 1.8-2.6 μM)