Learn More
MSC Intertape Aluminum Tape, 2.5 in x 60 yd, 4.6 mil
Catalog No. 501149819
Encompass
This aluminum foil tape features a 4.6 mil dead soft aluminum foil with an aggressive, long-lasting pressure-sensitive acrylic adhesive. It is engineered for general purpose seaming, patching, and sealing, enhancing appearance and maintaining vapor barrier integrity. This tape provides long-lasting adhesion to clean, dry surfaces.
  • Flame retardant and high shear adhesion
  • UV and mold resistant
  • Excellent reflective properties (heat, light)
  • Superior bonding when exposed to either sub zero or elevated temperatures
  • Release coated kraft liner
Catalog No. 50-114-9819 Supplier MSC Supplier No. 62472584
This aluminum foil tape features a 4.6 mil dead soft aluminum foil with an aggressive, long-lasting pressure-sensitive acrylic adhesive. It is engineered for general purpose seaming, patching, and sealing, enhancing appearance and maintaining vapor barrier integrity. This tape provides long-lasting adhesion to clean, dry surfaces.
  • Flame retardant and high shear adhesion
  • UV and mold resistant
  • Excellent reflective properties (heat, light)
  • Superior bonding when exposed to either sub zero or elevated temperatures
  • Release coated kraft liner