Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Zinc, Quantofix™ Test Strips

Catalog No. AA46651QA
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA46651QA 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
H314; Causes severe skin burns and eye damage. P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a; Do not breathe dust/fume/gas/mist/vapours/spray. IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. IF SWALLOWED: Rinse mouth. Do NOT induce vomiting. Store locked up. Dispose of contents/container in accordance with local/regional/national/international regulations.
Catalog No. AA46651QA Supplier Thermo Scientific Chemicals Supplier No. 046651QA

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

missing translation for 'validMainframeMsg'

H314; Causes severe skin burns and eye damage. P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a; Do not breathe dust/fume/gas/mist/vapours/spray. IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. IF SWALLOWED: Rinse mouth. Do NOT induce vomiting. Store locked up. Dispose of contents/container in accordance with local/regional/national/international regulations.

 

Zinc, Quantofix™ Test Strips is used to measure the determination of zinc in solutions quickly. It can be used readily for measurements. It is used to regularly check the plating baths in the metal plating industry. It is also used to check the concentration of zinc in cooling water treatment, where zinc salts are used as corrosion inhibitors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Liquid
  • Colorless
  • Fully miscible
TRUSTED_SUSTAINABILITY

Specifications

Format Liquid
For Use With (Application) Reagents and 100 strips to measure 0-100ppm Zn+2 in aqueous solution
Includes Reagents and 100 strips
Quantity 1 Kit
Type Fully Miscible

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.