Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Amberlite™ IRA-67, ion exchange resin

Catalog No. AA42253A1
Click to view available options
Quantity:
250 g
1 kg
5 kg

Acrylic-DVB; Weakly basic gel type resin | CAS: 80747-90-6 | Styrene-DVB

Amberlite™ IRA-67 is used for de-acidification and desalination of water and for isolation of acidic natural products. It allows for effective adsorption and desorption of naturally occurring organic molecules such as humic and fulvic acids that are present in many water supplies.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 80747-90-6
MDL Number MFCD00145567

Specifications

Physical Form Beads
Odor Odorless
Molecular Formula Styrene-DVB
Density 1.1-1.4g/cm3
Color Amber
Quantity 1 kg
Solubility Information Insoluble in water.
Chemical Name or Material Amberlite IRA-200C(Na), Ion Exchange Resin
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.