Amberlite IRN-150, Ion Exchange Resin, Alfa Aesar

Styrene-DVB; Mix of strongly acidic and basic gel type resin

Manufacturer: Alfa Aesar 40992A9

Quantity 10kg

 View more versions of this product

Catalog No. AA40992A9

  Check Availability
Add to Cart

    Chemical Identifiers

    Physical Form Beads
    Molecular Formula Styrene-DVB
    MDL Number MFCD00145822
    Chemical Name or Material Amberlite IRN-150

    Specifications

    Quantity 10kg
    Color Amber
    Density 1.1-1.3g/cm3
    Physical Form Beads
    Odor Odorless
    Molecular Formula Styrene-DVB
    MDL Number MFCD00145822
    Appearance Amber beads
    Ignition Point 500°C (932°F)
    Chemical Name or Material Amberlite IRN-150

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit