Alfa Aesar™ HPLC Grade 5 Micron APS Silica Gel

Manufacturer: Alfa Aesar 4580914

 View more versions of this product

Catalog No. AA4580914

  Check Availability
Add to Cart

    Specifications

    EINECS Number 231-545-4
    Product Type Silica gel
    Grade HPLC
    Quantity 25g
    Surface Area (Metric) 500m2/g
    Material Silica
    Particle Size
    Pore Size 70Å

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit