Learn More
Apexbio Technology LLC Alfacalcidol, 100mg. Cas: 41294-56-8 MFCD: MFCD00868328. Bone resorption inhibitor
SDP
Supplier:  Apexbio Technology LLC B2153100
Encompass_Preferred
Alfacalcidol is an analogue of Vitamin D and a potent inhibitor of bone resorption [1]. Alfacalcidol has been revealed to play a part in bone and mineral homeostasis via binding to the VDR (vitamin D receptor) in calcium-related target organs, such as intestine, bone, kidney and parathyroid gland. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5318