Learn More
Enzo Life Sciences Alfacalcidol (1mg). CAS: 41294-56-8
SDP
Supplier:  Enzo Life Sciences ALX460036M001
Encompass_Preferred
Converts to active calcitriol (1α,25-dihydroxyvitamin D3; BML-DM200) in vivo. Inhibits bone resorption. Alternative name: Hydroxyvitamin D3, 1-Hydroxycholecalciferol, (1alpha,3beta,5Z,7E)-9,10-Secocholesta-5,7,10(19)-triene-1,3-diol. Purity: ≥97% (Assay). Solubility: Soluble in 100% ethanol. Long Term Storage: -20°C.
Catalog No. 50-200-9292