Learn More
Medchemexpress LLC HY-10003 5mg Medchemexpress, Alfacalcidol CAS:41294-56-8 Purity:>98%
SDP
Catalog No. 501931701
Encompass_Preferred
Medchemexpress, HY-10003 5mg Alfacalcidol CAS:41294-56-8 Alfacalcidol (1-hydroxycholecalciferol) is a vitamin D active metabolites, acts as a non-selective VDR activator medication, and widely be used in the management of osteoporosis [1] [2] [3] [4] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-1701 Supplier Medchemexpress LLC Supplier No. HY100035MG
Add to Cart
Add to Cart
Medchemexpress, HY-10003 5mg Alfacalcidol CAS:41294-56-8 Alfacalcidol (1-hydroxycholecalciferol) is a vitamin D active metabolites, acts as a non-selective VDR activator medication, and widely be used in the management of osteoporosis [1] [2] [3] [4] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.