Learn More
Medchemexpress LLC Alfacalcidol-d6 | 1641940-94-4 | C27H38D6O2 | 5 MG
SDP
Catalog No. 5000373970
Encompass_Preferred
Alfacalcidol-d6 is the deuterium labeled Alfacalcidol, functioning as a non-selective VDR activator. This compound can be utilized as a tracer and an internal standard for quantitative analysis methods such as NMR, GC-MS, or LC-MS.
  • Deuterium-labeled compound
  • Non-selective VDR activator
  • Suitable for use as a tracer
  • Functions as an internal standard for quantitative analysis
  • Applicable in NMR, GC-MS, and LC-MS analytical techniques
Storage conditions: Powder: -20°C for 3 years, 4°C for 2 years. In solvent: -80°C for 2 years, -20°C for 1 year.
Catalog No. 50-003-73970 Supplier Medchemexpress LLC Supplier No. HY153325MG
Alfacalcidol-d6 is the deuterium labeled Alfacalcidol, functioning as a non-selective VDR activator. This compound can be utilized as a tracer and an internal standard for quantitative analysis methods such as NMR, GC-MS, or LC-MS.
  • Deuterium-labeled compound
  • Non-selective VDR activator
  • Suitable for use as a tracer
  • Functions as an internal standard for quantitative analysis
  • Applicable in NMR, GC-MS, and LC-MS analytical techniques
Storage conditions: Powder: -20°C for 3 years, 4°C for 2 years. In solvent: -80°C for 2 years, -20°C for 1 year.