Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfacalcidol 95.0+%, TCI America™
SDP

Catalog No. A312550MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A312550MG 50 mg
A312510MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A312550MG Supplier TCI America Supplier No. A312550MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 41294-56-8
Molecular Formula C27H44O2
Molecular Weight (g/mol) 400.647
MDL Number MFCD00868328
InChI Key OFHCOWSQAMBJIW-AQVMIQFASA-N
Synonym alphacalcidol
PubChem CID 87091364
IUPAC Name (1R,3R,5Z)-5-[2-[(1R,7aR)-7a-methyl-1-[(2R)-6-methylheptan-2-yl]-2,3,3a,5,6,7-hexahydro-1H-inden-4-ylidene]ethylidene]-4-methylidenecyclohexane-1,3-diol
SMILES CC(C)CCCC(C)C1CCC2C1(CCCC2=CC=C3CC(CC(C3=C)O)O)C

Specifications

Melting Point 136°C
Color White-Yellow
Quantity 50 mg
UN Number 2811
Formula Weight 400.65
Percent Purity ≥95.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Alfacalcidol
TSCA Yes
Recommended Storage Freezer (-20 °C)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.