Learn More
Cayman Chemical Alflutinib (mesylate), Purity: ≥98%, CAS: 2130958-55-1, 50 mg
SDP
Catalog No. 502992103
Encompass_Preferred
An inhibitor of mutant EGFRs; selective for EGFRL858R, EGFRG719X, EGFRL858R, EGFRL861A, and EGFRT790M mutant EGFRs over wild-type EGFR; reduces tumor growth in an EGFRL858R and EGFRT790M-expressing LU1868 NSCLC PDX mouse model at 10 and 30 mg/kg
Catalog No. 50-299-2103 Supplier Cayman Chemical Supplier No. 3147350 MG
An inhibitor of mutant EGFRs; selective for EGFRL858R, EGFRG719X, EGFRL858R, EGFRL861A, and EGFRT790M mutant EGFRs over wild-type EGFR; reduces tumor growth in an EGFRL858R and EGFRT790M-expressing LU1868 NSCLC PDX mouse model at 10 and 30 mg/kg