Learn More
Medchemexpress LLC Alfuzosin-d3 | 1006724-55-5 | 99.9% | 1 MG
SDP
Catalog No. 5000405684
Encompass_Preferred
Alfuzosin-d3 is deuterium labeled Alfuzosin. This compound can be used as a tracer and as an internal standard for quantitative analysis. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Stable heavy isotopes incorporated into drug molecules
  • Potential to affect pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-004-05684 Supplier Medchemexpress LLC Supplier No. HYB0192S21MG
Alfuzosin-d3 is deuterium labeled Alfuzosin. This compound can be used as a tracer and as an internal standard for quantitative analysis. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Stable heavy isotopes incorporated into drug molecules
  • Potential to affect pharmacokinetic and metabolic profiles of drugs