Learn More
Medchemexpress LLC Alfuzosin (hydrochloride) | 81403-68-1 | 99.7% | 100 MG
SDP
Catalog No. 5000406639
Encompass_Preferred
Alfuzosin hydrochloride is an orally active, selective, and competitive α1-adrenoceptor antagonist. It functions by relaxing the muscles of the prostate and bladder neck, thereby aiding in urination. This compound is utilized in research studies focusing on benign prostatic hyperplasia (BPH).
  • Orally active compound
  • Selective α1-adrenoceptor antagonist
  • Relaxes prostate and bladder neck muscles
  • Aids in urination
  • Useful in benign prostatic hyperplasia (BPH) studies
Catalog No. 50-004-06639 Supplier Medchemexpress LLC Supplier No. HYB0192A100MG
Alfuzosin hydrochloride is an orally active, selective, and competitive α1-adrenoceptor antagonist. It functions by relaxing the muscles of the prostate and bladder neck, thereby aiding in urination. This compound is utilized in research studies focusing on benign prostatic hyperplasia (BPH).
  • Orally active compound
  • Selective α1-adrenoceptor antagonist
  • Relaxes prostate and bladder neck muscles
  • Aids in urination
  • Useful in benign prostatic hyperplasia (BPH) studies