Learn More
Medchemexpress LLC Alfuzosin (hydrochloride) | 81403-68-1 | 99.7% | 25 MG
SDP
Catalog No. 5000451030
Encompass_Preferred
Alfuzosin hydrochloride is an orally active, selective, and competitive α1-adrenoceptor antagonist. It relaxes the muscles of the prostate and bladder neck, aiding in urination. This compound is used in the study of benign prostatic hyperplasia (BPH).
  • Orally active compound
  • Selective and competitive α1-adrenoceptor antagonist
  • Relaxes prostate and bladder neck muscles
  • Aids in urination
  • Used for benign prostatic hyperplasia (BPH) studies
Catalog No. 50-004-51030 Supplier Medchemexpress LLC Supplier No. 25MGHYB0192A
Alfuzosin hydrochloride is an orally active, selective, and competitive α1-adrenoceptor antagonist. It relaxes the muscles of the prostate and bladder neck, aiding in urination. This compound is used in the study of benign prostatic hyperplasia (BPH).
  • Orally active compound
  • Selective and competitive α1-adrenoceptor antagonist
  • Relaxes prostate and bladder neck muscles
  • Aids in urination
  • Used for benign prostatic hyperplasia (BPH) studies