Learn More
Medchemexpress LLC Alfuzosin hydrochloride | 81403-68-1 | 99.7% | 50 MG
SDP
Catalog No. 5000407391
Encompass_Preferred
Alfuzosin hydrochloride is an orally active, selective and competitive α1-adrenoceptor antagonist. It relaxes the muscles of the prostate and bladder neck, aiding in urination. This compound can be used in the study of benign prostatic hyperplasia (BPH).
  • Orally active
  • Selective and competitive α1-adrenoceptor antagonist
  • Relaxes the muscles of the prostate and bladder neck
  • Aids in urination
  • Can be used in the study of benign prostatic hyperplasia (BPH)
Catalog No. 50-004-07391 Supplier Medchemexpress LLC Supplier No. HYB0192A50MG
Alfuzosin hydrochloride is an orally active, selective and competitive α1-adrenoceptor antagonist. It relaxes the muscles of the prostate and bladder neck, aiding in urination. This compound can be used in the study of benign prostatic hyperplasia (BPH).
  • Orally active
  • Selective and competitive α1-adrenoceptor antagonist
  • Relaxes the muscles of the prostate and bladder neck
  • Aids in urination
  • Can be used in the study of benign prostatic hyperplasia (BPH)