Learn More
Cayman Chemical Alfuzosin (hydrochloride), Purity: ≥98%, CAS: 81403-68-1, 5 mg
SDP
Catalog No. 502955404
Encompass_Preferred
A post-synaptic α1-adrenergic receptor antagonist commonly used to improve lower urinary tract symptoms associated with benign prostatic hyperplasia; displays high-affinity with non-selectivity for the three known human α1 adrenoceptors (pKi = 8.0, 8.0, and 8.5 for α1A, α1B, and α1D, respectively)
Catalog No. 50-295-5404 Supplier Cayman Chemical Supplier No. 136485 MG
A post-synaptic α1-adrenergic receptor antagonist commonly used to improve lower urinary tract symptoms associated with benign prostatic hyperplasia; displays high-affinity with non-selectivity for the three known human α1 adrenoceptors (pKi = 8.0, 8.0, and 8.5 for α1A, α1B, and α1D, respectively)