Learn More
Aviva Systems Biology Corp ALG1 Antibody - N-terminal region (ARP49445_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP49445_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ALG1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ALG1. Alternate names: HMAT1, HMT-1, HMT1, MT-1, CDG1K, Mat-1, hMat-1ApplicationWB. Species reactivity: Cow, Human, Mouse, Rabbit, Rat, Yeast
Catalog No. 50-014-60088