Learn More
Aviva Systems Biology Corp ALG2 Antibody - C-terminal region (ARP48974_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP48974_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ALG2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human ALG2. Alternate names: CDGIi, FLJ14511, hALPG2, NET38ApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-014-59981